missing translation for 'onlineSavingsMsg'
Learn More

Boron ICP Standard, 10,000 ppm B in H2O, Ricca Chemical

Supplier:  Ricca Chemical Company PB10KW500

 View more versions of this product

Catalog No. PB10KW-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Boron ICP Standard
10043-35-3,7732-18-5,1336-21-6
10,000ppm B
1 mL = 10 mg B
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
Natural Poly Bottle
92.0,5.26,0.92
96.0,5.47,0.96
BH3O3
B(O)(O)O
61.831
CHEBI:33118

Chemical Identifiers

10043-35-3
61.831
7628
boric acid
BH3O3
KGBXLFKZBHKPEV-UHFFFAOYSA-N
CHEBI:33118
B(O)(O)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions