missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron ICP Standard, 10,000 ppm B in H2O, Ricca Chemical
Chemical Identifiers
| CAS | 10043-35-3 |
|---|---|
| Molecular Formula | BH3O3 |
| Molecular Weight (g/mol) | 61.831 |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| PubChem CID | 7628 |
| ChEBI | CHEBI:33118 |
| IUPAC Name | boric acid |
| SMILES | B(O)(O)O |
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|
PB10KW-50
|
Ricca Chemical Company
PB10KW50 |
Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
PB10KW-100
|
Ricca Chemical Company
PB10KW100 |
Natural LDPE Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
PB10KW-500
|
Ricca Chemical Company
PB10KW500 |
Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 10043-35-3 | |
| 61.831 | |
| 7628 | |
| boric acid |
| BH3O3 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| CHEBI:33118 | |
| B(O)(O)O |
Specifications
| Natural Poly Bottle | |
| 92.0,5.26,0.92 | |
| 96.0,5.47,0.96 | |
| BH3O3 | |
| B(O)(O)O | |
| 61.831 | |
| CHEBI:33118 |
| 10043-35-3,7732-18-5,1336-21-6 | |
| 10,000ppm B | |
| 1 mL = 10 mg B | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 7628 | |
| Boron ICP Standard |