missing translation for 'onlineSavingsMsg'
Learn More

Boron, AAS solution standard, Specpure™ B 1000μg/mL

H3BO3 in H2O

$75.55 - $75.55

Chemical Identifiers

Molecular Formula H3BO3 in H2O
MDL Number MFCD00134034
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA88055AE
View Documents
Thermo Scientific Chemicals
088055AE
100 mL
Each for $75.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

H3BO3 in H2O
MFCD00134034

Specifications

Liquid
MFCD00134034
Specpure
H3BO3 in H2O
100 mL
Boron, AAS standard solution, B 1000μg/mL
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H360FD

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only