missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borneol (contains ca. 20% Isoborneol) 70.0+%, TCI America™
$65.46 - $275.30
Chemical Identifiers
| CAS | 507-70-0 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.253 |
| MDL Number | MFCD00066426 |
| InChI Key | DTGKSKDOIYIVQL-HHCGNCNQSA-N |
| PubChem CID | 44630107 |
| IUPAC Name | (1R,3S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol |
| SMILES | CC1(C2CCC1(C(C2)O)C)C |
Chemical Identifiers
| 507-70-0 | |
| 154.253 | |
| DTGKSKDOIYIVQL-HHCGNCNQSA-N | |
| (1R,3S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol |
| C10H18O | |
| MFCD00066426 | |
| 44630107 | |
| CC1(C2CCC1(C(C2)O)C)C |
Specifications
| 507-70-0 | |
| White | |
| MFCD00066426 | |
| 1312 | |
| CC1(C2CCC1(C(C2)O)C)C | |
| 154.253 | |
| 154.25 | |
| Crystalline Powder |
| 201°C | |
| C10H18O | |
| 25 g | |
| DTGKSKDOIYIVQL-HHCGNCNQSA-N | |
| (1R,3S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol | |
| 44630107 | |
| ≥70.0% (GC) | |
| Borneol (contains ca. 20% Isoborneol) |
Safety and Handling
EINECSNumber : (4)-0607
RTECSNumber : ED7000000
TSCA : Yes