accessibility menu, dialog, popup

UserName

CAS RN 507-70-0

H3CH3CCH3OH

CAS RN 507-70-0

IUPAC Name: 1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Synonyms: baros bhimsaim camphor camphane, 2-hydroxy- 2-hydroxy-camphane malayan camphor bornyl alcohol 2-camphanol borneo camphor dryobalanops camphor 2-hydroxycamphane
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: DTGKSKDOIYIVQL-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(O)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results