missing translation for 'onlineSavingsMsg'
Learn More

(-)-Borneol, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111020215
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020215 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020215 Supplier Merck Emd Millipore Supplier No. 155981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(-)-Borneol is an enantiomer. It is a bicyclic monoterpene compound used gengrally for analgesia and anaesthesia. It is considered as positive modulators of GABA receptors.

Featured Industries: Food and Beverages

Specifications

CAS 464-45-9
Flash Point 65°C
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00003759
Synonym endo-(1 S)-1,7,7-Trimethylbicyclo[2.2.1 ]heptan-2-ol
Type Analytical Standard
Quantity 1 g
Formula Weight 154.25
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less