missing translation for 'onlineSavingsMsg'
Learn More

(-)-Borneol, 97+%

CAS: 464-45-9 | C10H18O | 154.25 g/mol

Supplier:  Thermo Scientific Chemicals A126840B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA126840B


Only null left

Description

Description

(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.

Solubility
Soluble in chloroform, ethanol, acetone, ether, benzene, toluene, decalin and tetralin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

464-45-9
154.25
DTGKSKDOIYIVQL-NQMVMOMDSA-N
1201518
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
C10H18O
MFCD00003759,MFCD00066426,MFCD00066427
--borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
CHEBI:15394

Specifications

(-)-Borneol
208°C to 211°C
65°C (149°F)
C10H18O
MFCD00003759,MFCD00066426,MFCD00066427
3587558
--borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
DTGKSKDOIYIVQL-NQMVMOMDSA-N
−36° (c=5 in Ethanol)
154.25
CHEBI:15394
≥97%
464-45-9
210°C
97+%
1000 g
UN1312
14,1338
Soluble in chloroform,ethanol,acetone,ether,benzene,toluene,decalin and tetralin. Insoluble in water.
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
(1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
1201518
154.25
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q

H228-H315-H319-H500

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: BORNEOL

EINECSNumber : 207-353-1

RTECSNumber : DT5095000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions