Learn More
(-)-Borneol, 97+%
Description
(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.
Solubility
Soluble in chloroform, ethanol, acetone, ether, benzene, toluene, decalin and tetralin. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 208°C to 211°C |
| Boiling Point | 210°C |
| Flash Point | 65°C (149°F) |
| Assay Percent Range | 97+% |
| Quantity | 50 g |
| UN Number | UN1312 |
| Beilstein | 3587558 |
| Merck Index | 14,1338 |
| Solubility Information | Soluble in chloroform,ethanol,acetone,ether,benzene,toluene,decalin and tetralin. Insoluble in water. |
| IUPAC Name | (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol |
| Show More |
Safety and Handling
| Hazard Category | H228-H315-H319-H500 |
| Hazard Statement | GHS H Statement H228 |
| Precautionary Statement | P210-P240-P241-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: BORNEOL |
| EINECSNumber | 207-353-1 |
| RTECSNumber | DT5095000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only