missing translation for 'onlineSavingsMsg'
Learn More

Boric Anhydride, Purified, 97%, Spectrum™ Chemical

Catalog No. p-7165152
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609187 125 g Poly Bottle
18609189 500 g Poly Bottle
18609186 2.5 kg Poly Bottle
18609188 12 kg Poly Pail
18609190 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609187 Supplier Spectrum Chemical Mfg Cor Supplier No. B1135125GM
Only null left

Chemical Identifiers

CAS 1303-86-2

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 97%
Grade Purified
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.