missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Boric Acid with Sher Indicator, pH 4.65
Description
- Ready to use boric acid
- Maintains the correct concentration, pH, and indicator ratio
- For pH or colorimetric titration
- pH adjusted to 4.65 ± 0.15
- For pH or colorimetric titration
Spécifications
Spécifications
| Chemical Name or Material | Boric Acid with Sher Indicator |
| CAS | 64-17-5, 1310-73-2, 10043-35-3, 7732-18-5, 76-60-8 |
| Quantity | 5 L |
| Molecular Formula | BH3O3 |
| Packaging | Plastic Bottle |
| Synonym | Orthoboric Acid |
| Molecular Weight (g/mol) | 61.83 |
| Concentration or Composition (by Analyte or Components) | 0.02 |