missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%, Solstice

Catalog No. 6002311
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002311 500 g
6002309 1 kg
6002310 2.5 kg
60045920 6 x 1 kg
60045921 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6002311 Supplier Solstice Supplier No. 31146500G

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Vapor Pressure 2.6mmHg (20°C)
Quantity 500 g
Linear Formula H3BO3
UN Number NONH for all modes of transport
Formula Weight 61.83g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Boric Acid, Reag. Ph. Eur. reag. ISO
Hazard Statement P201-P280-P308 + P313
Precautionary Statement H360FD
EINECSNumber 233-139-2
RTECSNumber ED4550000