missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Purified, Reagents

Catalog No. p-200021077
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337609 2.5 kg
01337608 500 g
01337607 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337609 Supplier Labchem Inc Supplier No. C21043002.5A5
Only null left

Boric Acid, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless/White
Melting Point 168°C to 170°C
Boiling Point 300°C
Packaging Poly Bottle
Specific Gravity 1.435
Quantity 2.5 kg
Physical Form Solid
Grade Pure
Chemical Name or Material Boric Acid, Purified