Learn More
Boric acid, Puratronic™, 99.9995% (metals basis)
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Supplier: Thermo Scientific Chemicals 01065922
Description
It is used as a primary or adjunct buffer in swimming pools, flame retardant, neutron absorber in nuclear power plants, colorant to make flame green, preservative in urine samples, welding flux in combination with borax, or precursor to other chemical compounds. It is used to prevent/remove existing wet and dry rot in timbers, treat wood against fungal/insect attack, prevent athleteâs foot, lubricate carom boards, and reduce surface oxidation on metals at high temperatures. It can be used to produce lubricant for ceramic or metal surfaces, textile fiberglass, induction furnace linings and ceramics, Silly Putty, and glass LCD flat panel displays. It can prevent the high temperature reaction of aluminum with nitrates in pyrotechniques, and neutralize active hydrofluoric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Boric Acid | |
| 10043-35-3 | |
| None | |
| 100 g | |
| BH3O3 | |
| H3BO3 | |
| 1697939 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 7628 | |
| 61.83 | |
| Puratronic |
| ∽185°C (decomposition) | |
| 1.435 | |
| 99.99% (metals basis) | |
| (metals basis) | |
| Powder | |
| MFCD00011337 | |
| 14,1336 | |
| Soluble in water- solubility increases rapidly with temperature. Soluble in methanol and ethanol. Very slightly soluble in acetone. | |
| OB(O)O | |
| 61.83 | |
| CHEBI:33118 | |
| 99.9995% |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| 7628 | |
| boric acid |
| BH3O3 | |
| MFCD00011337 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| CHEBI:33118 | |
| OB(O)O |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P260-P281-P308+P313-P501c
H360FD
EINECSNumber : 233-139-2
RTECSNumber : ED4550000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only