missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Powder, Technical, Spectrum™ Chemical

Catalog No. 18609185
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609185 500 g Poly Pail
18609183 125 g Poly Pail
18609179 2.5 kg Poly Pail
18609184 12 kg Poly Pail
18609180 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609185 Supplier Spectrum Chemical Mfg Cor Supplier No. B1122500GM
Only null left

Chemical Identifiers

CAS 10043-35-3
Molecular Weight (g/mol) 61.83

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 500 g
Grade Technical
Chemical Name or Material Boric Acid

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.