missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Powder, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18609176
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609176 125 g Poly Bottle
18609178 500 g Poly Bottle
18609181 2.5 kg Poly Pail
18609177 12 kg Poly Pail
18609182 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609176 Supplier Spectrum Chemical Mfg Cor Supplier No. B1125125GM

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 1
Heavy Metals 1e-05
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99.5%
Grade ACS

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.