missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Molecular Biology Grade (orthoboric acid), Promega™
Description
- Quality tested and certified free of DNase, RNase and protease activity
Specifications
Specifications
| CAS | 10043-35-3 |
| Assay Percent Range | ≥99.5% |
| Quantity | 1kg |
| Molecular Formula | BH3O3 |
| Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| SMILES | B(O)(O)O |
| IUPAC Name | boric acid |
| Molecular Weight (g/mol) | 61.831 |
| PubChem CID | 7628 |
| Show More |
Safety and Handling
| Recommended Storage | Store at 15° to 30°C |