missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Boric acid, Molecular Biology Grade, 99.5% min

Catalog No. p-7057842
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67419A1 1 kg
AAJ67419A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAJ67419A1 Supplier Thermo Scientific Chemicals Supplier No. J67419A1
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

Used to promote amidations of carboxylic acids and amines in catalytic amounts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point ∼185°C (decomposition)
Density 1.435 g/mL
Odor Odorless
Quantity 1 kg
Physical Form Powder
Beilstein 1697939
Merck Index 14,1336
Formula Weight 61.83
Percent Purity ≥99.5%
Grade Molecular Biology
Chemical Name or Material Boric acid
Show More Show Less
Hazard Category H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 233-139-2
RTECSNumber ED4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only