missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Granular, Technical, Spectrum™ Chemical

Catalog No. 18609174
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609174 2.5 kg Fiber Drum
18609175 500 g Fiber Drum
18609172 100 lb. Fiber Drum
18609173 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609174 Supplier Spectrum Chemical Mfg Cor Supplier No. B11332.5KG
Only null left

Chemical Identifiers

CAS 10043-35-3
Molecular Weight (g/mol) 61.83

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 2.5 kg
Grade Technical
Chemical Name or Material Boric Acid

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.