missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Granular, NF, EP, BP, JP, 99.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Boric Acid, Granular, NF, EP, BP, JP is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| pH | 3.8-4.8 |
| CAS Max % | 100% |
| Packaging | Poly Bottle |
| Quantity | 125 g |
| Percent Purity | 99.5 to 100.5% |
| Grade | Multi-Compendial |