missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-31202
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A73500 500 g Poly Bottle
A731 1 kg Poly Bottle
A733 3 kg Poly Bottle
A7310 10 kg Poly Pail
A7350 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A73500 Supplier Thermo Fisher Scientific Supplier No. FLA73500
Edge

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point 169°C
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Non_volatile Matter 0.05% max.
Packaging Poly Bottle
Vapor Pressure 2.7mbar at 20°C
Assay Percent Range ≥99.5 %
Quantity 500 g
Physical Form Powder/Solid
Formula Weight 61.83
Percent Purity ≥99.5%
Grade Certified ACS
Chemical Name or Material Boric Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:0
Instability:1