missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Crystal, NF, 99.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Boric Acid, Crystal, NF is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Pail |
| Quantity | 2.5 kg |
| Percent Purity | 99.5 to 100.5% |
| Grade | NF |