missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Crystal, NF, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18601040
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601040 125 g Poly Bottle
18601041 500 g Poly Bottle
18601042 2.5 kg Poly Pail
18601043 12 kg Poly Pail
18601044 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601040 Supplier Spectrum Chemical Mfg Cor Supplier No. BO112125GM
Only null left

BO112, 10043-35-3, H3BO3

Spectrum™ Chemical Boric Acid, Crystal, NF is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99.5 to 100.5%
Grade NF