missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. p-7215407
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCB0500500 500 g
RDCB0500100 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCB0500500 Supplier Ricca Chemical Company Supplier No. RDCB0500500B1
Only null left

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging White Poly Wide Mouth Bottle
Quantity 500 g
Percent Purity ≥ 99.5%
Grade ACS
Chemical Name or Material Boric Acid
ShelfLife 60 months