missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCB0500100
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCB0500100 1 kg
RDCB0500500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCB0500100 Supplier Ricca Chemical Company Supplier No. RDCB05001000B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 100
Heavy Metals (as Pb) max. 0.001%
Non_volatile Matter max. 0.05%
Packaging White Poly Wide Mouth Bottle
Quantity 1 kg
Percent Purity min 99.5%
Grade ACS
Chemical Name or Material Boric Acid
ShelfLife 84 months