missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.99% (metals basis)

Catalog No. p-3256933
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3677136 500 g
AA36771A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3677136 Supplier Thermo Scientific Chemicals Supplier No. 03677136
Only null left

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

It is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is too employed in leather manufacture, electroplating, and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Melting Point ∽185°C (decomposition)
Density 1.43515
Flash Point None
Assay Percent Range 99.99% (metals basis)
Quantity 500 g
Physical Form &Plus;8 Mesh, Granular
Beilstein 1697939
Solubility Information It is fairly soluble in boiling water but less in cold water. Soluble in lower alcohols,acetone and moderately soluble in pyridine.
Formula Weight 61.83
Chemical Name or Material Boric acid
Hazard Category H360FD
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 233-139-2
RTECSNumber ED4550000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only