missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. p-3734218
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327132500 250 g
AC327130010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327132500 Supplier Thermo Scientific Chemicals Supplier No. 327132500
Only null left

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer, Lewis acid

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color White
Melting Point 169.0°C
Heavy Metals (as Pb) 50ppm max.
Assay Percent Range 99.5%
Quantity 250 g
Physical Form Powder
Linear Formula H3BO3
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Solubility Information Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
Formula Weight 61.83
Percent Purity 99.5%
Grade Molecular Biology
Chemical Name or Material Boric acid, DNAse, RNAse and Protease Free
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only