missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 99.5% (ACS Guaranteed Reagent), MilliporeSigma™

Catalog No. MBX08651
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBX08651 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBX08651 Supplier Technidata Supplier No. BX08651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Quantity 500 g
Physical Form Crystalline
Formula Weight 61.83
Grade ACS
Chemical Name or Material Boric Acid, 99.5%