missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 98%

Catalog No. AAA166240I
Change view
Click to view available options
Quantity:
5000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA166240I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA166240I Supplier Thermo Scientific Chemicals Supplier No. A166240I

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

Solubility
Soluble in glycerol, ether, alcohol, methanol, liquid ammonia, acetone (slightly), water (6.35 g/100 cc) at 30°C, water (27.6 g/100 cc) at 100°C, water (50 g/L) at 21°C, and water (1 g / 18 ml cold water).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Melting Point ∼185°C (decomposition)
Density 1.435
Odor Odorless
Flash Point None
Linear Formula H3BO3
Quantity 5000 g
Beilstein 1697939
Merck Index 14,1336
Solubility Information Soluble in glycerol,ether,alcohol,methanol,liquid ammonia,acetone (slightly),water (6.35g/100 cc) at 30°C,water (27.6g/100 cc) at 100°C,water (50g/L) at 21°C,and water (1 g / 18 ml cold water).
Formula Weight 61.83
Percent Purity 98%
Chemical Name or Material Boric acid
Show More Show Less
Hazard Category H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 233-139-2
RTECSNumber ED4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only