missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, 4% w/v with Methyl Red-Bromocresol Green Indicators, Reagents
Specifications
Specifications
| Chemical Name or Material | Boric Acid, 4% w/v with Methyl Red-Bromocresol Green Indicators |
| pH | 4.5 to 5 |
| Color | Purple |
| CAS | 7732-18-5, 10043-35-3 |
| Boiling Point | 100°C |
| Packaging | Cubitainer |
| Specific Gravity | 1.015 |
| Quantity | 20 L |
| Physical Form | Liquid |
| Percent Purity | 4% |
| Show More |