missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, 2% w/v, without Indicator, Reagents
Boric Acid, 2% w/v, without Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CB11240020F
Specifications
| 4.5 to 5 | |
| Colorless | |
| 100°C | |
| 1.0074 | |
| Liquid | |
| Reagent |
| Boric Acid, 2% w/v, without Indicator | |
| 7732-18-5,10043-35-3 | |
| Cubitainer | |
| 20 L | |
| 2% |