missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 2% w/v, without Indicator, Reagents

Catalog No. 01335779
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335779 1 L
01335778 20 L
01335777 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335779 Supplier Labchem Inc Supplier No. CB1124001A

Boric Acid, 2% w/v, without Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Boric Acid, 2% w/v, without Indicator
pH 4.5 to 5
Color Colorless
CAS 7732-18-5, 10043-35-3
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0074
Quantity 1 L
Physical Form Liquid
Percent Purity 2%
Grade Reagent
Show More Show Less