missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 2% w/v with Methyl Red-Methylene Blue Indicators, Reagents

Catalog No. p-200021966
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336576 20 L
01336575 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336576 Supplier Labchem Inc Supplier No. C710421020F
Only null left

Boric Acid, 2% w/v with Methyl Red-Methylene Blue Indicators, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Boric Acid, 2% w/v with Methyl Red-Methylene Blue Indicators
pH 4.5 to 5
Color Burgundy
CAS 7732-18-5, 10043-35-3
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0074
Quantity 20 L
Physical Form Liquid
Percent Purity 2%
Grade Reagent
Show More Show Less