missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 2% w/v with Methyl Red-Methylene Blue Indicators, Reagents

Catalog No. 01336578
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336578 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336578 Supplier Labchem Inc Supplier No. C710420020F
Only null left

Boric Acid, 2% w/v with Methyl Red-Methylene Blue Indicators, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Boric Acid, 2% w/v with Methyl Red-Methylene Blue Indicators
pH 4.5 to 5
Color Burgundy
CAS 7732-18-5, 10043-35-3
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0074
Quantity 20 L
Physical Form Liquid
Percent Purity 2%
Grade Reagent
Show More Show Less