missing translation for 'onlineSavingsMsg'
Learn More

Bordeaux Red, TCI America™

Catalog No. B077925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B077925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B077925G Supplier TCI America Supplier No. B077925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5858-33-3
Molecular Formula C20H12N2Na2O7S2
Molecular Weight (g/mol) 502.423
MDL Number MFCD00003887
InChI Key MRVNLKITQIBNKU-LSHDGIFOSA-L
Synonym acid red 17, acid bordeaux, azo bordeaux, bordeaux b, bordeaux r, acid bordeaux a, acid bordeaux b, java bordeaux b, bordeaux red b, azo bordeaux ss
PubChem CID 9570215
IUPAC Name disodium;(4Z)-4-(naphthalen-1-ylhydrazinylidene)-3-oxonaphthalene-2,7-disulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Purple-Red
Quantity 25 g
Formula Weight 502.42
Physical Form Crystalline Powder
Chemical Name or Material Bordeaux Red
RTECSNumber QJ6479500
TSCA Yes