accessibility menu, dialog, popup

UserName

CAS RN 5858-33-3

IUPAC Name: disodium (4Z)-4-[2-(naphthalen-1-yl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (4Z)-4-[2-(naphthalen-1-yl)hydrazin-1-ylidene]-3-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 502.42
Molecular Formula: C20H12N2Na2O7S2
InChi Key: MRVNLKITQIBNKU-LSHDGIFOSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N/NC1=C2C=CC=CC2=CC=C1)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results