missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Borate Buffer |
| CAS Min % | 97, 0.46, 0.07 |
| CAS Max % | 100, 0.48, 0.07 |
| Color | Colorless |
| CAS | 1303-96-4, 1310-73-2 |
| For Use With (Application) | Nitrogen Analysis |
| pH | 9.5 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Packaging | Natural Poly Bottle |
| Show More |