missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical
For Analysis of Ammonia and organic Nitrogen
$198.94 - $199.66
Specifications
| Chemical Name or Material | Borate Buffer |
|---|---|
| Color | Colorless |
| Name Note | pH 9.5 |
| CAS | 7732-18-5,1303-96-4,1310-73-2 |
| CAS Min % | 97.0,0.46,0.07 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
1040-32
|
Ricca Chemical Company
RI104032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
R10400004F
|
Ricca Chemical Company
R10400004F |
4 L | Cubitainer |
N/A
|
|
||||
Specifications
| Borate Buffer | |
| pH 9.5 | |
| 97.0,0.46,0.07 | |
| Laboratory | |
| Non-sterile |
| Colorless | |
| 7732-18-5,1303-96-4,1310-73-2 | |
| 100.0,0.48,0.07 | |
| 9.5 |