missing translation for 'onlineSavingsMsg'
Learn More

Borate Buffer, pH 10.00, Ricca Chemical

Catalog No. R2105100500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2105100500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2105100500 Supplier Ricca Chemical Company Supplier No. R2105100500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Borate Buffer
CAS 7732-18-5, 7447-40-7, 10043-35-3, 1310-73-2
Color Colorless Liquid
CAS Max % 99.15, 0.37, 0.31, 0.17
pH 10
Quantity 500 mL
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Laboratory