missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borate Buffer, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Borate Buffer |
| Name Note | For Nitrogen (Ammonia), Nitrogen (Organic) |
| CAS | 7732-18-5, 1303-96-4, 1310-73-2 |
| Color | Colorless |
| CAS Max % | 99.48, 0.48, 0.04 |
| Decomposition Information | Boron |
| pH | 9.50 ±0.20 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Identification | Passes Test |
| Show More |