missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.5

Catalog No. p-7042329
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62154AK 250 mL
AAJ62154AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62154AK Supplier Thermo Scientific Chemicals Supplier No. J62154AK

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

Solubility
Soluble in water.

Notes
Store at 25°C. This product is relatively stable under this condition. Best result obtained with use within 6-8 months of purchase.

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 9.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water.
Chemical Name or Material Borate buffer soln.
Hazard Category H319-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only