Learn More
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.0
CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol
Supplier: Thermo Scientific Chemicals J62125AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Borate buffer soln. | |
| 0.5 M | |
| Liquid | |
| B4Na2O7 | |
| NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
| disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
| 1330-43-4 | |
| 9.0 | |
| 250 mL | |
| MFCD00081185 | |
| [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 | |
| 201.21 |
Chemical Identifiers
| 1330-43-4 | |
| 201.21 | |
| NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
| [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
| B4Na2O7 | |
| MFCD00081185 | |
| disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H360FD
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only