missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 8.5

Catalog No. AAJ60803AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60803AP 500 mL
AAJ60803AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60803AP Supplier Thermo Scientific Chemicals Supplier No. J60803AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. BBS has many uses because it is isotonic and has a strong bactericidal effect. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer. Finfs uses in Protein-coating on microtiter plates, Blocking solution in blots together with Casein or BSA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. BBS has many uses because it is isotonic and has a strong bactericidal effect. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer. Finfs uses in Protein-coating on microtiter plates, Blocking solution in blots together with Casein or BSA.

Solubility
Soluble in water 26 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 8.5
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in water 26g/L (20°C).
Chemical Name or Material Borate buffer soln.
Hazard Category H319-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only