missing translation for 'onlineSavingsMsg'
Learn More

Borane - Pyridine Complex 80.0+%, TCI America™

Catalog No. B156925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B156925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B156925ML Supplier TCI America Supplier No. B156925ML

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridine Borane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1

Specifications

Boiling Point 65°C
Quantity 25 mL
UN Number 1992
Formula Weight 92.94
Percent Purity ≥80.0% (T)
Physical Form Liquid
Chemical Name or Material Borane - Pyridine Complex
RTECSNumber US3675000
TSCA Yes
Recommended Storage Refrigerator