accessibility menu, dialog, popup

UserName

BH3N

CAS RN 110-51-0

IUPAC Name: (pyridin-1-ium-1-yl)boranuide
Synonyms: pyridin-1-ium-1-ylboranuide
Molecular Weight (g/mol): 92.94
Molecular Formula: C5H8BN
InChi Key: LPGWNCNRGQANGC-UHFFFAOYSA-N
SMILES: [BH3-][N+]1=CC=CC=C1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)