missing translation for 'onlineSavingsMsg'
Learn More

Borane - Morpholine Complex 95.0+%, TCI America™

Catalog No. M08985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08985G 5 g
M089825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08985G Supplier TCI America Supplier No. M08985G

Chemical Identifiers

CAS 4856-95-5
Molecular Formula C4H12BNO
Molecular Weight (g/mol) 100.96
MDL Number MFCD00066736
InChI Key IFVAHVOGOBRFSP-UHFFFAOYSA-N
Synonym morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine
PubChem CID 6327109
IUPAC Name morpholine borane
SMILES B.C1COCCN1

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
UN Number 2921
Formula Weight 100.96
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Borane - Morpholine Complex
RTECSNumber QD8530000
TSCA Yes