missing translation for 'onlineSavingsMsg'
Learn More

Borane - Ammonia Complex 90.0+%, TCI America™

Catalog No. B50825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50825G 5 g
B508225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50825G Supplier TCI America Supplier No. B50825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13774-81-7
Molecular Formula BH3N
Molecular Weight (g/mol) 27.841
MDL Number MFCD00075639
InChI Key JBANFLSTOJPTFW-UHFFFAOYSA-N
Synonym Ammonia Borane
PubChem CID 6332567
IUPAC Name azane;boron
SMILES [B].N

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 30.87
Percent Purity ≥90.0% (V)
Physical Form Crystalline Powder
Chemical Name or Material Borane - Ammonia Complex
TSCA No
Recommended Storage Refrigerator