accessibility menu, dialog, popup

UserName

BNH3

CAS RN 13774-81-7

IUPAC Name: amine boron
Synonyms: ammonia borane
Molecular Weight (g/mol): 27.84
Molecular Formula: BH3N
InChi Key: JBANFLSTOJPTFW-UHFFFAOYSA-N
SMILES: [B].N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results