missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ BoosterExpress™ Reagents and Kit
Description
- Significantly enhanced gene expression levels in diverse cell lines
- Compatibility with all major commercial transfection reagents including GenePORTER™ Transfection Reagents
- Optimized conditions for popular cell lines
- Efficient for both transient and stable transfections
- Fast and easy protocol
- Set of chemical cocktails designed to significantly increase gene expression levels
Specifications
Specifications
| Concentration | 100X |
| For Use With (Application) | Designed to significantly increase gene expression levels, efficient for both transient and stable transfections |
| Quantity | 3 x 1.5 mL |
| Reagent Type | Reagent 3 |