missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Empty Mini Gel Cassettes

Catalog No. NW2010
Encompass
Change view
Click to view available options
Quantity:
20 cassettes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NW2010 20 cassettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NW2010 Supplier Invitrogen™ Supplier No. NW2010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Allows users to pour custom gel formulations while still enjoying the advantages of the Mini Gel Tank

Bolt Empty Gel Cassettes give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank and XCell SureLock Mini-Cell. The cassettes are sealed on three sides (so no gel leakage) and feature the unique wedge-shaped well. They are available in the standard 1-mm thickness and can be used with Bolt™ 10- or 12-well combs (Cat. No. NW3010 and NW3012). Since Bolt™ empty cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary.

Specifications

For Use With (Equipment) Mini Gel Tank and XCell SureLock Mini-Cell
Gel Size Mini (8 x 8 cm)
Product Line Bolt
Type Empty Mini Gel Cassette
Quantity 20 cassettes
Shipping Condition Room Temperature
Do you offer empty cassettes and combs for casting Mini and Midi protein gels?

We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock Mini-Cell, Mini Gel Tank, or XCell4 SureLock Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/power-supplies-protein-electrophoresis.html#staining).

- The Empty Mini Gel Cassettes are available in two thicknesses (1.0 mm and 1.5 mm) and can be used with Invitrogen Combs that are available in two thicknesses (1.0 mm and 1.5 mm), and in 7 types (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
- The Bolt Empty Mini Gel Cassettes are available in 1.0 mm thickness and can be used with Bolt 10- or 12-well combs.
- The Empty Midi Gel Cassettes are available in 1.0 mm thickness and can be used with three Invitrogen Comb types (12 + 2 wells, 20 wells, and 26 wells).

For Research Use Only. Not for use in diagnostic procedures.