missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Boldenone Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175001
Description
Working Concentration: 0.125 μg/mL. Sensitivity: 10 ng/mL Alpha boldonone produces 94% inhibition in a competitive ELISA employing 17αBoldenone polyclonal antibody.
Boldenone is an anabolic steroid developed for veterinary use, mostly for treatment of horses.Specifications
| Boldenone | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Unconjugated | |
| C19H26O2 | |
| 17a-Boldenone(3)-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |